Merck
Influence of supports on photocatalytic degradation of phenol and 4-chlorophenol in aqueous suspensions of titanium dioxide.
Journal of environmental sciences (China) (2013-04-20)
Kashif Naeem, Feng Ouyang
ABSTRAKT
The photocatalytic degradation of phenol and 4-chlorophenol (4-CP) in aqueous suspensions with the use of titanium dioxide (TiO2) under UV irradiation was examined. The effects of different supporting materials mixed physically with TiO2 were studied to achieve maximum degradation efficiency. Among the three supports, namely activated carbon (AC), silica (SiO2) and zeolite (ZSM-5), all exhibited paramount efficiency for degradation of phenol and 4-CP and was better than TiO2 alone. The optimum concentration was found to be 50 mg for all supporting materials. The efficiency order of the three supports was as follows: AC > ZSM-5 > SiO2, respectively. Whilst, the degradation of phenol and 4-CP was improved from 70.6% to 87.6% and 80.6% to 89.7%, respectively, within 120 min photocatalysis in the presence of optimal amount of AC. The degradation was also comparatively enhanced in the presence of cheaper rice husk and the activity was closed to ZSM-5 and lower than AC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Chlorophenol, ≥99%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material