- Case of sorafenib-induced thyroid storm.
Case of sorafenib-induced thyroid storm.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-04-24)
Sigurdis Haraldsdottir, Quan Li, Miguel A Villalona-Calero, Thomas E Olencki, Kari Kendra, Steven W Ing
PMID23610115
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Nicotinamide, European Pharmacopoeia (EP) Reference Standard