Merck
Stalling the engine of resistance: targeting cancer metabolism to overcome therapeutic resistance.
Cancer research (2013-04-24)
Ethan B Butler, Yuhua Zhao, Cristina Muñoz-Pinedo, Jianrong Lu, Ming Tan
ABSTRAKT
Cancer cells are markedly different from normal cells with regards to how their metabolic pathways are used to fuel cellular growth and survival. Two basic metabolites that exemplify these differences through increased uptake and altered metabolic usage are glucose and glutamine. These molecules can be catabolized to manufacture many of the building blocks required for active cell growth and proliferation. The alterations in the metabolic pathways necessary to sustain this growth have been linked to therapeutic resistance, a trait that is correlated with poor patient outcomes. By targeting the metabolic pathways that import, catabolize, and synthesize essential cellular components, drug-resistant cancer cells can often be resensitized to anticancer treatments. The specificity and efficacy of agents directed at the unique aspects of cancer metabolism are expected to be high; and may, when in used in combination with more traditional therapeutics, present a pathway to surmount resistance within tumors that no longer respond to current forms of treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland