Merck
Phase 2 study of azacytidine plus sorafenib in patients with acute myeloid leukemia and FLT-3 internal tandem duplication mutation.
Blood (2013-04-25)
Farhad Ravandi, Mona Lisa Alattar, Michael R Grunwald, Michelle A Rudek, Trivikram Rajkhowa, Mary Ann Richie, Sherry Pierce, Naval Daver, Guillermo Garcia-Manero, Stefan Faderl, Aziz Nazha, Marina Konopleva, Gautam Borthakur, Jan Burger, Tapan Kadia, Sara Dellasala, Michael Andreeff, Jorge Cortes, Hagop Kantarjian, Mark Levis
ABSTRAKT
Patients received 5-azacytidine (AZA) 75 mg/m(2) intravenously daily for 7 days and sorafenib 400 mg orally twice daily continuously; cycles were repeated at ~1-month intervals. Forty-three acute myeloid leukemia (AML) patients with a median age of 64 years (range, 24-87 years) were enrolled; 37 were evaluable for response. FMS-like tyrosine kinase-3 (FLT3)-internal tandem duplication (ITD) mutation was detected in 40 (93%) patients, with a median allelic ratio of 0.32 (range, 0.009-0.93). They had received a median of 2 prior treatment regimens (range, 0-7); 9 had failed prior therapy with a FLT3 kinase inhibitor. The response rate was 46%, including 10 (27%) complete response with incomplete count recovery (CRi), 6 (16%) complete responses (CR), and 1 (3%) partial response. The median time to achieve CR/CRi was 2 cycles (range, 1-4), and the median duration of CR/CRi was 2.3 months (range, 1-14.3 months). Sixty-four percent of patients achieved adequate (defined as >85%) FLT3 inhibition during their first cycle of therapy. The degree of FLT3 inhibition correlated with plasma sorafenib concentrations. FLT3 ligand levels did not rise to levels seen in prior studies of patients receiving cytotoxic chemotherapy. The combination of AZA and sorafenib is effective for patients with relapsed AML and FLT-3-ITD. This trial was registered at clinicaltrials.gov as #NCT01254890.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
5-Azacytidine, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®