Merck
Dehydration of glycerol over niobia-supported silicotungstic acid catalysts.
Journal of nanoscience and nanotechnology (2013-05-08)
Young Yi Lee, Hye Jeong Ok, Dong Ju Moon, Jong Ho Kim, Nam Cook Park, Young Chul Kim
ABSTRAKT
Liquid-phase dehydration of glycerol to acrolein over nanosized niobia-supported silicotungstic acid catalysts was performed to investigate the effect of the silicotungstic acid loading on the catalytic performance of the catalysts. The catalysts were prepared by following an impregnation method with different HSiW loadings in the range of 10-50 wt%. The prepared catalysts were characterized by N2 physisorption, XRD, FT-IR, TPD of ammonia, and TGA. Dehydration of glycerol was conducted in an autoclave reactor under the conditions of controlled reaction temperatures under corresponding pressure. Increasing HSiW loading rapidly increased the acidity of HSiW/Nb205 catalyst and rate of glycerol conversion, but acrolein selectivity decreased due to enhanced deactivation of the catalyst by carbon deposit. Consequently, it was confirmed that catalytic activity for the dehydration of glycerol to acrolein was dependant on the acidity of catalyst and can be controlled by HSiW loading.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis