Merck
  • Crystal structures of the catalytic domain of a novel glycohydrolase family 23 chitinase from Ralstonia sp. A-471 reveals a unique arrangement of the catalytic residues for inverting chitin hydrolysis.
Crystal structures of the catalytic domain of a novel glycohydrolase family 23 chitinase from Ralstonia sp. A-471 reveals a unique arrangement of the catalytic residues for inverting chitin hydrolysis.
The Journal of biological chemistry (2013-05-10)
Takao Arimori, Noriko Kawamoto, Shoko Shinya, Nobuo Okazaki, Masami Nakazawa, Kazutaka Miyatake, Tamo Fukamizo, Mitsuhiro Ueda, Taro Tamada
ABSTRAKT
Chitinase C from Ralstonia sp. A-471 (Ra-ChiC) has a catalytic domain sequence similar to goose-type (G-type) lysozymes and, unlike other chitinases, belongs to glycohydrolase (GH) family 23. Using NMR spectroscopy, however, Ra-ChiC was found to interact only with the chitin dimer but not with the peptidoglycan fragment. Here we report the crystal structures of wild-type, E141Q, and E162Q of the catalytic domain of Ra-ChiC with or without chitin oligosaccharides. Ra-ChiC has a substrate-binding site including a tunnel-shaped cavity, which determines the substrate specificity. Mutation analyses based on this structural information indicated that a highly conserved Glu-141 acts as a catalytic acid, and that Asp-226 located at the roof of the tunnel activates a water molecule as a catalytic base. The unique arrangement of the catalytic residues makes a clear contrast to the other GH23 members and also to inverting GH19 chitinases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chitin from shrimp shells, BioReagent, suitable for analysis of chitinase, purified powder
Sigma-Aldrich
Chitin from shrimp shells, practical grade, powder