Merck
Intranasal substituted cathinone "bath salts" psychosis potentially exacerbated by diphenhydramine.
Journal of addiction medicine (2013-06-05)
Erik W Gunderson, Matthew G Kirkpatrick, Laura M Willing, Christopher P Holstege
ABSTRAKT
In this report, we describe a case of intranasal "bath salts"-associated psychosis. Symptoms developed during a 3-week binge and were potentially exacerbated by oral diphenhydramine taken for insomnia. The clinical case conference includes expert discussion from 3 disciplines: emergency medicine toxicology, behavioral pharmacology, and addiction medicine. It is hoped that the discussion will provide insight into the clinical aspects and challenges of addressing acute substituted cathinone toxicity, including acute psychosis, a major adverse effect of bath salts consumption.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
R(+)-Cathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Diphenhydramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Diphenhydramine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard