Merck
Adsorption equilibria and kinetics for the adsorption of p-nitrophenol on a phenoxy groups modified hypercrosslinked polystyrene resin.
Journal of nanoscience and nanotechnology (2013-06-13)
Xiaomei Wang, Chenxi Zhao, Jianhan Huang
ABSTRAKT
Adsorption equilibria and kinetics of p-nitrophenol adsorbed on HJ-01 were investigated in this study. The result indicated that phenoxy groups were uploaded on the skeleton of HJ-01 successfully, the Friedel-Crafts reaction brought on prodigious changes for the Brunauer-Emmett-Teller (BET) specific surface area and pore structure while the nucleophilic substitution reaction had few effects. The adsorption experiments revealed that the acidic solution was suitable for p-nitrophenol adsorption, the adsorption isotherms could be characterized by the Freundlich isotherm equation and the adsorption thermodynamic parameters were all negative, the adsorption kinetic curves obeyed the pseudo-second-order rate equation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material