Merck
  • Uterine artery pulsatility index improves prediction of methotrexate resistance in women with gestational trophoblastic neoplasia with FIGO score 5-6.
Uterine artery pulsatility index improves prediction of methotrexate resistance in women with gestational trophoblastic neoplasia with FIGO score 5-6.
BJOG : an international journal of obstetrics and gynaecology (2013-06-14)
A Sita-Lumsden, H Medani, R Fisher, R Harvey, D Short, N Sebire, P Savage, A Lim, M J Seckl, R Agarwal
ABSTRAKT
The Uterine Artery Pulsatility Index (UAPI) is an ultrasound measure of tumour vascularity. In this study, we hypothesised that a UAPI ≤ 1 (high vascularity) would identify women with gestational trophoblastic neoplasia (GTN) at increased risk of resistance to first-line single-agent methotrexate (MTX-R). Single-centre cohort study. Charing Cross Hospital, a UK national centre for the treatment of trophoblastic disease. All women with a GTN FIGO score 5-6 treated with methotrexate (n = 92), between 1999 and 2011, at Charing Cross Hospital. UAPI was measured before the start of chemotherapy, and women were monitored for the development of MTX-R. Frequency of MTX-R in women with UAPI ≤ 1 compared with UAPI >1. UAPI was measured before chemotherapy in 73 of 92 women with GTN FIGO score 5-6. UAPI ≤ 1 predicted MTX-R independent of the FIGO score (hazard ratio 2.9, P = 0.04), with an absolute risk of MTX-R in women with a UAPI ≤ 1 of 67% (95% CI 53-79%) compared with 42% (95% CI 24-61%) with a UAPI >1 (P = 0.036). Our results suggest UAPI is an independent predictor of MTX-R in women with FIGO 5-6 GTN.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®