Merck
[Affinity capture of specific DNA fragments with the use of short synthetic sequences].
Bioorganicheskaia khimiia (2013-07-13)
V S Mikhaĭlov, V K Potapov, R N Amirkhanov, N V Amirkhanov, S S Bulanenkova, S B Akopov, V F Zarytova, L G Nikolaev, E D Sverdlov
ABSTRAKT
The ability of short peptide nucleic acid (PNA) oligomers and oligonucleotides containing modified residues of 5-methylcitidine, 2-aminoadenosine and 5-propynyl-2'-deoxyuridine (strong binding oligonucleotides, SBO) to affinity capture the target double-stranded DNA fragment from mixture by means of the end invasion was compared. Both types of probes were highly effective at the conditions used. The SBO-based probes may represent a handy and easily prepared alternative to PNA for selection of target DNA fragments from mixtures.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material