Merck
Thiohistidine biosynthesis.
Chimia (2013-07-19)
Florian P Seebeck
ABSTRAKT
Ergothioneine and ovothiol A are sulfur-containing histidine derivatives produced by microorganisms including Mycobacterium tuberculosis, Trypanosoma cruzi or Erwinia amylovora and may also play important roles in human physiology. Based on our recent identification of thiohistidine biosynthetic enzymes from Mycobacterium smegmatis and Erwinia tasmaniensis we investigate several aspects of sulfur-based redox biochemistry. For example, we are characterizing the catalytic mechanism of two thiohistidine biosynthetic enzymes which afford O2-dependent sulfur insertion into the C(5)-H and C(2)-H bonds of the imidazolyl side chain of histidine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Safrole, ≥97%
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
Hydroquinidine, 95%
Sigma-Aldrich
Selenic acid solution, 40 wt. % in H2O, 99.95% trace metals basis