Merck
Kinetics of solvents release from paint coatings. I. Paint coating hardened at +20 degrees C temperature.
Bulletin of the Institute of Maritime and Tropical Medicine in Gdynia (1986-01-01)
R Wiglusz, E Gałuszko, I Jarnuszkiewicz, G Delag
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
1,2,3-Trimethylbenzene, analytical standard