Merck
More on fructosamine assay as index to glycated protein.
Clinical chemistry (1986-10-01)
R N Johnson, J R Baker
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Deoxy-1-morpholino-D-fructose, ≥98% (TLC)