Merck
Determination of synthetic food dyes by capillary electrophoresis.
Journal of chromatography. A (1994-10-07)
S Suzuki, M Shirao, M Aizawa, H Nakazawa, K Sasa, H Sasagawa
ABSTRAKT
A method for the determination of synthetic tar dyes used as food additives using capillary electrophoresis with photodiode-array detection was investigated. The dyes Erythrosine (R-3), Phloxine (R-104), Rose Bengal (R-105), Acid Red (R-106), Amaranth (R-2), New Coccine (R-102) and Allura Red AC (R-40) were separated on a capillary column (50 cm x 75 microns I.D.) and identified from the absorbance spectra of each peak. The electrophoresis buffer used was a mixture of 25 mM sodium phosphate buffer and 25 mM sodium borate buffer (1:1) (pH 8.0) containing 10 mM sodium dodecyl sulfate (SDS). Substitution of beta-cyclodextrin for SDS in the electrolyte buffer was effective for the separation of R-2 and R-102. This modified method could be employed as an additional assay method for these two dyes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ponceau 4R, analytical standard
Supelco
Allura Red AC, analytical standard
Sigma-Aldrich
Allura Red AC, Dye content 80 %
Sigma-Aldrich
New Coccine, Dye content 75 %