- Fixed drug eruption caused by sulfaguanidine.
Fixed drug eruption caused by sulfaguanidine.
Dermatology (Basel, Switzerland) (1994-01-01)
J Arias, M Fernández-Rivas, M Martinez, C Senent
PMID8075454
ABSTRAKT
We report a patient who developed a fixed drug eruption (FDE) due to sulfaguanidine. This drug has not been previously implicated as a cause of FDE. The diagnosis was confirmed by an oral provocation and a biopsy of the lesion. Oral provocations with other sulfonamides, sulfamethoxazole and sulfadiazine, were well tolerated.