Merck
Factors affecting the production of carbon monoxide in inhalation anesthetic use.
Anesthesia and analgesia (1996-06-01)
C M Callan
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Barium hydroxide, technical grade, ~95%
Sigma-Aldrich
Barium hydroxide solution, 0.3 N
Sigma-Aldrich
Barium hydroxide octahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Barium hydroxide monohydrate, 98%
Sigma-Aldrich
Barium hydroxide octahydrate, ≥98%