Merck
TEST yolk buffer and antisperm antibodies--interaction?
Fertility and sterility (1997-03-01)
D Katsoff, J H Check
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TES, ≥99% (titration)