Merck
beta-Glucuronidase in common duct bile, methodological aspects, variation of pH optima and relation to gallstones.
Scandinavian journal of clinical and laboratory investigation (1997-07-01)
T Osnes, O Sandstad, V Skar, M Osnes
ABSTRAKT
beta-Glucuronidase of human or bacterial origin may deconjugate bilirubin diglucuronide, causing pigment gallstones. Intrinsic interference by biliary compounds must be minimized for accurate assay of beta-glucuronidase. We report a modified ion-pair extraction of interfering substances by tetrahexylammonium chloride (THAC) in ethyl acetate in the presence of albumin, and a microtitre plate assay for biliary beta-glucuronidase activity in bile with the substrate p-nitrophenol-glucuronide. Adding albumin improved the recovery of beta-glucuronidase activity to 99.8% (CV 1.9%), and 92.2% of the bilirubin in bile samples was extracted in one step. Competitive inhibition was overcome by increasing the substrate concentration. In endoscopically obtained common duct bile from 44 patients, five different beta-glucuronidase activity peaks were identified, at pH 3.9, 4.8, 5.3, 5.8 and 7.2. The pH profiles were classified into one bacterial pattern and five patterns for presumed human beta-glucuronidase. Of the latter patterns, four displayed dual activity peaks. In a second sample, obtained at follow up in four patients, their original pH profile was maintained. In conclusion, using the modified purification and assay system, we found functionally diverse subcategories of human beta-glucuronidase with respect to activity at variable pH. Our results indicate that several pH optima have to be taken into consideration in order to clarify the role of human biliary beta-glucuronidase in the pathogenesis of pigment gallstones. Bacterial beta-glucuronidase activity was associated with duodenal diverticula (p < 0.05) and common duct stones (p < 0.05).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahexylammonium hydrogensulfate, ≥98.0% (T)
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Supelco
Tetrahexylammonium hydrogensulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetrahexylammonium chloride, 96%
Sigma-Aldrich
Tetrahexylammonium hydroxide solution, ~40% in H2O (T)
Sigma-Aldrich
Tetrahexylammonium bromide, 99%
Supelco
Tetrahexylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)