Merck
Torsade de pointes after pipamperone intoxication.
Pharmacy world & science : PWS (1998-06-10)
L Bont, H A Bosker, F Brus, J P Yska, F H Bosch
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pipamperone dihydrochloride, ~99% (HPLC), powder