Merck
The metalloprotease meprin β generates amino terminal-truncated amyloid β peptide species.
The Journal of biological chemistry (2012-08-11)
Jessica Bien, Tamara Jefferson, Mirsada Causević, Thorsten Jumpertz, Lisa Munter, Gerd Multhaup, Sascha Weggen, Christoph Becker-Pauly, Claus U Pietrzik
ABSTRAKT
The amyloid β (Aβ) peptide, which is abundantly found in the brains of patients suffering from Alzheimer disease, is central in the pathogenesis of this disease. Therefore, to understand the processing of the amyloid precursor protein (APP) is of critical importance. Recently, we demonstrated that the metalloprotease meprin β cleaves APP and liberates soluble N-terminal APP (N-APP) fragments. In this work, we present evidence that meprin β can also process APP in a manner reminiscent of β-secretase. We identified cleavage sites of meprin β in the amyloid β sequence of the wild type and Swedish mutant of APP at positions p1 and p2, thereby generating Aβ variants starting at the first or second amino acid residue. We observed even higher kinetic values for meprin β than BACE1 for both the wild type and the Swedish mutant APP form. This enzymatic activity of meprin β on APP and Aβ generation was also observed in the absence of BACE1/2 activity using a β-secretase inhibitor and BACE knock-out cells, indicating that meprin β acts independently of β-secretase.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Actinonin