Merck
You're the flight surgeon: lymphatic filariasis.
Aviation, space, and environmental medicine (2013-03-02)
Manoj Ravi
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Albendazole, analytical standard, ≥98%
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Albendazole, European Pharmacopoeia (EP) Reference Standard