Merck
Drug safety: double jeopardy.
Nature (2013-06-28)
Mary Carmichael
MATERIAŁY
Numer produktu
Marka
Opis produktu
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)