Merck
India's health ministry bans pioglitazone, metamizole, and flupentixol-melitracen.
BMJ (Clinical research ed.) (2013-07-09)
Soumyadeep Bhaumik
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pioglitazone hydrochloride, ≥98% (HPLC)