Merck
Development of immunoassays for tyramine and tryptamine toxins of Phalaris aquatica L.
Journal of agricultural and food chemistry (2000-01-19)
J H Skerritt, S L Guihot, S E McDonald, R A Culvenor
ABSTRAKT
The leaves of the perennial pasture grass Phalaris aquatica L. (phalaris) contain two groups of known toxins, indole alkaloids, primarily dimethyltryptamines and N-methyltyramines, which cause illnesses in grazing animals, especially sheep. Using amino-reactive and phenolic hydroxyl-reactive homobifunctional reagents, simple methods were devised for coupling toxins representative of those in phalaris to carrier proteins and enzymes for ELISA development. ELISAs were produced for both groups of toxins. Dimethyltryptamines were most sensitively detected [lower limit of detection (LLD) of 1 microg/L for bufotenine] using rabbit anti-bufotenine antibodies, coupled to ovalbumin using divinyl sulfone, with detection using a peroxidase conjugate prepared using the same hapten coupled with 1, 4-butanediol diglycidyl ether. The assay cross-reacted with other toxins of the same class (N,N-dimethyltryptamine and N, N-dimethyl-5-methoxytryptamine) but not with the structurally related amino acids histidine and tryptophan. The most sensitive N-methyltyramine assay (LLD of 1 microg/mL for N-methyltyramine) utilized antisera to tyramine with N-methyltyramine coupled to peroxidase. Significant cross-reaction was seen with the low-grade toxin hordenine, but detection of tyramine was poorer, whereas the amino acid tyrosine was not detected. These assays could be applied to the analysis of simple extracts of Phalaris leaves with minimal interference. A good correspondence was observed between toxin levels by ELISA and estimates from a more tedious thin-layer chromatography method. The method has now been incorporated in a Phalaris breeding program.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hordenine, ≥97.0% (HPLC)