Merck
  • [The effect of oral ovulation inhibiting (mono-, bi- and triphase) contraceptives and of Continuin and Postinor on proteo- and steroid hormones].
[The effect of oral ovulation inhibiting (mono-, bi- and triphase) contraceptives and of Continuin and Postinor on proteo- and steroid hormones].
Acta pharmaceutica Hungarica (1992-01-01)
M Farkas
ABSTRAKT
Endocrinological changes due to the effect of treatment with mono-, bi- and three-phase contraceptive pills taking continuously and occasionally are reviewed. It is established that oral contraceptives based on ovulation inhibition are safe. Less side effects are produced by Ovidon tablets. Continuin and Postinor pills are not safe, and they can produce irregular bleeding. Beside their numerous advances extrauterine pregnancy frequently happens (6.4%). A three-phase preparation is expected to have a reversible effect and lack of a consequent functional infertility. This is improved by hormone level measurements.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethynodiol diacetate