- Extraction of 20(S)-ginsenoside Rg2 from cultured Panax notoginseng cells in vitro stimulates human umbilical cord vein endothelial cell proliferation.
Extraction of 20(S)-ginsenoside Rg2 from cultured Panax notoginseng cells in vitro stimulates human umbilical cord vein endothelial cell proliferation.
American journal of therapeutics (2006-06-15)
Xiujuan Xin, Jianwen Liu, Xiqiang Li, Jianjiang Zhong, Dongzhi Wei
PMID16772761
ABSTRAKT
From the cultured cells of Panax notoginseng, a kind of ginseng saponin-ginsenoside Rg2, was separated and purified, and its structure was elucidated as 6-O-[alpha-L-rhamnose (l-->2)-beta-D-glucopyranosyl]3beta, 12beta, 20(S)-trihydroxydammar-24-ene [20(S)-ginsenoside Rg2]. Moreover, its bioactivity stimulates human umbilical cord vein endothelial cell proliferation; the growth of the cells treated with 20(S)-ginsenoside Rg2 was 14-fold higher than that of untreated ones, and the level of the cell's plasminogen activator (PA) was also twofold higher.
MATERIAŁY