Merck
Development and characterization of methacrylate-based hydrazide monoliths for oriented immobilization of antibodies.
Journal of chromatography. A (2009-02-11)
P Brne, Y-P Lim, A Podgornik, M Barut, B Pihlar, A Strancar
ABSTRAKT
Convective interaction media (CIM; BIA Separations) monoliths are attractive stationary phases for use in affinity chromatography because they enable fast affinity binding, which is a consequence of convectively enhanced mass transport. This work focuses on the development of novel CIM hydrazide (HZ) monoliths for the oriented immobilization of antibodies. Adipic acid dihydrazide (AADH) was covalently bound to CIM epoxy monoliths to gain hydrazide groups on the monolith surface. Two different antibodies were afterwards immobilized to hydrazide functionalized monolithic columns and prepared columns were tested for their selectivity. One column was further tested for the dynamic binding capacity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adipic acid dihydrazide, ≥98% (titration)