Merck
Intracellular pH regulation by Na⁺/H⁺ exchanger-1 (NHE1) is required for growth factor-induced mammary branching morphogenesis.
Developmental biology (2012-03-01)
Edmund C Jenkins, Shawon Debnath, Stephen Gundry, Sajini Gundry, Umit Uyar, Jimmie E Fata
ABSTRAKT
Regulation of intracellular pH (pHi) and protection against cytosolic acidification is primarily a function of the ubiquitous plasma membrane Na+/H+exchanger-1 (NHE1), which uses a highly conserved process to transfer cytosolic hydrogen ions (H+) across plasma membranes in exchange for extracellular sodium ions (Na+). Growth factors, which are essential regulators of morphogenesis, have also been found to be key activators of NHE1 exchanger activity; however, the crosstalk between both has not been fully evaluated during organ development. Here we report that mammary branching morphogenesis induced by transforming growth factor-alpha (TGFα) requires PI3K-dependent NHE1-activation and subsequent pHi alkalization. Inhibiting NHE1 activity after TGFα stimulation with 10 μM of the NHE1-specific inhibitor N-Methyl-N-isobutyl Amiloride (MIA) dramatically disrupted branching morphogenesis, induced extensive proliferation, ectopic expression of the epithelial hyper-proliferative marker Keratin-6 and sustained activation of MAPK. Together these findings indicate a novel developmental signaling cascade involving TGFα>PI3K>NHE1>pHi alkalization, which leads to a permissible environment for MAPK negative feedback inhibition and thus regulated mammary branching morphogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder