Merck
Evaluation of radiogallium-labeled, folate-embedded superparamagnetic nanoparticles in fibrosarcoma-bearing mice.
Journal of cancer research and therapeutics (2012-07-31)
Seyyedeh Leila Hosseini-Salekdeh, Amir Reza Jalilian, Hassan Yousefnia, Kammaledin Shafaii, Majid Pouladian, Morteza Mahmoudi
ABSTRAKT
Elevated expression of the folate receptor (FR) occurs in many human malignancies. Thus, folate targeting is widely utilized in drug delivery purposes specially using nano-radioactive agents. In this work, we report production and biological evaluation of gallium-67 labeled superparamagnetic iron oxide nanoparticles, embedded by folic acid ( 67 Ga-SPION-folate) complex especially in tumor-bearing mice for tumor imaging studies. The structure of SPION-folate was confirmed by X-ray diffraction (XRD), transmission electron microscopy (TEM) and foureir transform infrared spectroscopy (FT-IR) analyses. The radiolabeled SPION-folate formation was confirmed by instant thin layer chromatography (ITLC). Tumor induction was performed by the use of poly-aromatic hydrocarbon injection in rodents as reported previously. [ 67 Ga]-SPION-folate was shown to possess a particle size of ≈ 5-10 nm using instrumental methods followed by ITLC test. Biocompatibility of the compound was investigated using an 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay followed by stability tests and tumor accumulation studies in fibrosarcoma-bearing mice after subcutaneous (s.c.) application. All values were expressed as mean ± standard deviation (mean ± SD) and the data were compared using Student t-test. Statistical significance was defined as P<0.05. [ 67 Ga]-SPION-folate was prepared by a modified co-precipitation method possessing a particle size of ≈ 5-10 nm using instrumental methods (>95% radiochemical purity). Biodistribution studies demonstrated tumor:blood, tumor:bone and tumor:muscle ratios of 4.23, 4.98 and 11.54 respectively after 24 h. Due to the nano-scale size and high-penetrative property of the developed folate-containing nano-complex, this system can be an interesting drug delivery modality with therapeutic applications and folate receptor-targeting behavior, while possessing paramagnetic properties for thermotherapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
3-Methylcholanthrene solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
3-Methylcholanthrene, 98%
Supelco
3-Methylcholanthrene, analytical standard