- Editorial Commentary: Enterococcus faecalis infective endocarditis: is it time to abandon aminoglycosides?
Editorial Commentary: Enterococcus faecalis infective endocarditis: is it time to abandon aminoglycosides?
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2013-02-09)
Jose M Munita, Cesar A Arias, Barbara E Murray
PMID23392395