Merck
  • Real-time monitoring on the adsorption process of salicylic acid onto chitosan membrane using dielectric spectroscopy: macroscale concentration polarization and dynamics.
Real-time monitoring on the adsorption process of salicylic acid onto chitosan membrane using dielectric spectroscopy: macroscale concentration polarization and dynamics.
The journal of physical chemistry. B (2013-02-28)
Yuhong Li, Weidong Gao, Kongshuang Zhao, Gang Yang, Zhonghe Zhu, Rongjing Cui
ABSTRAKT
The adsorption process of salicylic acid (SA) onto chitosan membrane is monitored in real time by the dielectric relaxation spectroscopy (DRS) method. A unique dielectric relaxation, which is related to the macroscale concentration polarization layers (CPLs) in the SA solution caused by the adsorption, is observed. By modeling the measured systems composed of the membrane, the CPLs, and the SA solution, the dielectric spectra are analyzed systematically on the basis of the interfacial polarization theory. The parameters about the constituent phases, i.e., the dielectric constant εm and the conductivity κm of the chitosan membrane, the conductivity distribution (κ1 to κ2), and the thickness dCPL of the CPL, are obtained. The time-dependent εm and κm give insight into the microstate of the chitosan membrane during the adsorption. Furthermore, the time evolution of the conductivity gradient of the CPL, Δκ/dCPL, is combined to interpret the adsorption mechanism. It is suggested that the noninvasive dielectric monitoring may be applied to many adsorption and release processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Salicylic acid, ≥99%, FG