Merck
Phytochemical investigation of Annulohypoxylon ilanense, an endophytic fungus derived from Cinnamomum species.
Chemistry & biodiversity (2013-03-16)
Ming-Der Wu, Ming-Jen Cheng, Ih-Sheng Chen, Yung-Shun Su, Sung-Yuan Hsieh, Hsun-Shuo Chang, Chun-Wei Chang, Gwo-Fang Yuan
ABSTRAKT
Cultivation of the fungal strain Annulohypoxylon ilanense, an endophytic fungus isolated from the wood of medicinal plant Cinnamomum species, resulted in the isolation of one new furanoid derivative, ilanefuranone (1), one new pyrrole alkaloid, ilanepyrrolal (2), and one new biarylpropanoid derivative, ilanenoid (3), together with 22 known compounds, of which one α-tetralone analog, (-)-(4R)-3,4-dihydro-4,6-dihydroxynaphthalen-1(2H)-one (4) was isolated for the first time from a natural source. The structures were elucidated on the basis of physicochemical evidence, in-depth NMR spectroscopic analysis, and high-resolution mass spectrometry, and the antimycobacterial activities were also evaluated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG