Merck
Improving wet and dry strength properties of recycled old corrugated carton (OCC) pulp using various polymers.
Carbohydrate polymers (2013-04-03)
Yahya Hamzeh, Sanaz Sabbaghi, Alireza Ashori, Ali Abdulkhani, Farshid Soltani
ABSTRAKT
In this study, the application of different dosages of low and high molecular weights (MW) of chitosan (Ch), cationic starch (CS) and poly vinyl alcohol (PVA) were systematically investigated using old corrugated carton (OCC) furnishes. Various sequences of above-mentioned polymeric additives were also examined to find out the optimal combination for improving both wet and dry tensile strength. For each treatment, 4 handsheets, each having basis weight of 100 g/m(2), were made. In general, the tensile strength of handsheets was significantly affected by the addition of polymeric agents. The enhancing effect of additives on dry tensile property was much higher than wet condition. The results also showed that the tensile strength of the samples made from OCC furnishes were improved upon the addition of high molecular weight chitosan (ChI) compared to the untreated ones (control). The low MW chitosan did not change the properties of handsheets dramatically. Application of polymeric agents moderately decreased the stretch to rupture, however with increasing dosage the stretch was improved. Sequential addition of used polymers showed that triple application of polymers was beneficial to both dry and wet tensile strength, although the effect was larger for dry. The best results in wet and dry tensile strengths were achieved using sequential of PVA-ChI-CS. Sequential addition of oppositely charged polymers forms a macromolecular layered structure of polyelectrolytes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 150,000 (average), 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Starch from corn, practical grade