Merck
The treatment of cystinosis with cysteamine and phosphocysteamine in the United Kingdom and Eire.
Pediatric nephrology (Berlin, Germany) (1995-12-01)
W G van't Hoff, N Gretz
ABSTRAKT
Fifty-nine patients with cystinosis were treated with cysteamine or phosphocysteamine in the United Kingdom up to May 1990. Treatment was started at a median age of 3.2 years (range 0.6-24.8 years) and continued for a median duration of 3.0 years (range 0.01-1.2 years). At the end of the study, 46 (78%) patients remained on treatment. One patient developed end-stage renal failure and 6 died. Efficacy was assessed in the 44 pre-transplant patients. The United Kingdom pre-transplant patients had significantly lower plasma creatinine concentrations at 6 and 8 years than a historical group of patients who did not receive cysteamine (P < 0.0001 and P < 0.0003, respectively). There was no significant difference between pretreatment and final post-treatment height standard deviation scores, suggesting maintenance of growth rate. The leucocyte cystine concentration was less than the accepted upper limit of the treatment range (1 nmol 1/2 cystine/mg protein) in only 21% of determinations. There was no significant difference between the mean pre-treatment and final values of leucocyte cystine concentration. The mean final doses of cysteamine (33 mg/kg per day) and phosphocysteamine (37 mg/kg per day base equivalent) were less than the mean dose (51 mg/kg per day) used in a United States multicentre trial. We conclude that cysteamine treatment was beneficial, but further improvements might be achieved by an improvement in monitoring of therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cysteamine S-phosphate sodium salt, analytical standard