- Synthesis, DNA-damaging and cytotoxic properties of novel topoisomerase II-directed bisantrene analogues.
Synthesis, DNA-damaging and cytotoxic properties of novel topoisomerase II-directed bisantrene analogues.
Bioorganic & medicinal chemistry letters (1999-01-01)
G Zagotto, A Oliva, F Guano, E Menta, G Capranico, M Palumbo
PMID9871638
ABSTRAKT
New bisantrene analogues were synthesized, bearing one or two 4,5-dihydro-1H-imidazol-2-yl hydrazone side chains at positions 1,4 or 9 of the anthracene ring system. A 10-azabioisostere was also prepared. The position of substituents in structurally isomeric drugs modulates topoisomerase II poisoning and specificity, along with cytotoxicity.
MATERIAŁY