Merck
Altered gene expression in melanocytes exposed to 4-tertiary butyl phenol (4-TBP): upregulation of the A2b adenosine receptor 1.
The Journal of investigative dermatology (1999-11-26)
I C Le Poole, F Yang, T L Brown, J Cornelius, G F Babcock, P K Das, R E Boissy
ABSTRAKT
Exposure to phenolic agents contributes to the development of occupational vitiligo. Proposed as a causative factor for leukoderma in vivo, the para-substituted phenol 4-tertiary butyl phenol was chosen to investigate early cellular events responsible for selective disappearance of melanocytes from the epidermis of individuals sensitive to such agents. To this end, differential display of melanocyte mRNA isolated from three separate cultures was performed following a 12 h exposure of cells to 250 microM 4-tertiary butyl phenol or to vehicle alone. Fragments of cDNA representing differentially expressed messages were cloned and subsequently confirmed by reverse dot blotting. Alignment analysis revealed that the L30 ribosomal protein was upregulated by the treatment, potentially reflecting altered levels of protein synthesis in response to stress. In addition, a gene sequence upregulated following exposure to 4-tertiary butyl phenol was identified as the A2b receptor (a P1 receptor for adenosine). Differential expression of this gene was confirmed in an RNase protection assay. By reverse transcription-polymerase chain reaction, the gene was shown to be expressed in keratinocytes and fibroblasts as well. Flow cytometry confirmed differential expression in melanocytes and fibroblasts, but not in keratinocytes. Interestingly, it has been reported that P1 purinoceptor stimulation can induce apoptosis. This is in concordance with results reported elsewhere demonstrating induction of apoptosis by 4-tertiary butyl phenol in human melanocytes, as well as with morphologic changes observed in this study in cells exposed to 250 microM 4-tertiary butyl phenol for 72 h. In conclusion, differential display is useful to establish melanocyte components involved in the cellular response to phenolic agents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)