Merck
Effect of biflavones of Ginkgo biloba against UVB-induced cytotoxicity in vitro.
The Journal of dermatology (2001-07-14)
S J Kim
ABSTRAKT
The effect of Ginkgo biloba extract on Ultraviolet B (UVB) irradiated fibroblasts was examined by using a neutral red dye uptake assay and a lactic dehydrogenase (LDH) release assay. Crude extract along with individual components, including flavone-glycosides and biflavones, were applied to cultured normal human skin fibroblasts for 12 hours, and 0, 20, 40 and 80 mJ/cm2 of UVB were irradiated. Two synthetic flavonoids, quercetin and rutin, which have polyphenol structures close to the flavonoids in Ginkgo biloba extract, were used to compare any structure-related activity under the same conditions. At the concentrations (from 0.25 to 2 mg/ml) treated with biflavone components (isoginkgetin/ginkgetin, sciadopitysin) and quercetin, high neutral red dye uptake was detected with gradual increases in UVB irradiation. The time-course release of LDH was determined as the cytotoxicity index (%) during 24 hours following a high dose UVB irradiation (200 mJ/cm2), and the pattern of this cytotoxicity index was similar to that of the neutral red dye uptake results. Sciadopitysin, isoginkgetin/ginkgetin and quercetin treatments lowered cytotoxicity indices to 50.81, 67.81 and 62.19%, respectively, compared to 95.38% for the untreated control. The antioxidant potential of biflavones of Ginkgo biloba could be explained on the basis of structure-related activity; hydroxy- and methyl-substitutions on the basic structure of these flavonoids played a role, as other reports have suggested.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Flavone, ≥99.0%