Merck
  • Rapid HPLC-electrospray tandem mass spectrometric assay for urinary testosterone and dihydrotestosterone glucuronides from patients with benign prostate hyperplasia.
Rapid HPLC-electrospray tandem mass spectrometric assay for urinary testosterone and dihydrotestosterone glucuronides from patients with benign prostate hyperplasia.
Clinical chemistry (2003-02-01)
Man Ho Choi, Jung Nyun Kim, Bong Chul Chung
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard