Merck
Branched-chain amino acid-enriched nutritional support in surgical and cancer patients.
The Journal of nutrition (2005-12-21)
Haroon A Choudry, Ming Pan, Anne M Karinch, Wiley W Souba
ABSTRAKT
Prolonged surgical stress and advanced malignant disease lead to systemic catabolism characterized by depletion of muscle protein and oxidation of skeletal muscle BCAA. BCAA oxidation provides energy for muscle and other organs and is the precursor for amino acid synthesis to replenish alanine and glutamine depleted in catabolic states. Persistent excessive catabolism leads to skeletal muscle wasting, negative nitrogen balance, and immune compromise. BCAAs, especially leucine, stimulate protein synthesis, inhibit proteolysis (in cell culture models and in animals), and promote glutamine synthesis. A number of small and diverse clinical trials studied the effects of BCAA-enriched nutritional support in moderately to severely stressed surgical and cancer patients. The findings of these clinical trials have been inconsistent; some show improved nitrogen balance, increased skeletal muscle protein synthesis, and reduced skeletal muscle catabolism whereas others show no significant improvement. The value of these trials is compromised by small sample size, heterogeneous patients, poor study design, varying degrees of metabolic stress, and inappropriate endpoints. More recent trials that evaluate clinical outcomes in hepatocellular carcinoma patients show promising results; in addition to improving metabolic parameters, BCAA-enriched oral supplementation improved morbidity and quality of life in patients undergoing major liver resection and chemo-embolization. In summary, the role of BCAAs in the nutritional support of stressed surgical and cancer patients remains to be clearly defined, despite their potential beneficial biological properties.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture