Merck
Trabulsiella guamensis, a new genus and species of the family Enterobacteriaceae that resembles Salmonella subgroups 4 and 5.
Journal of clinical microbiology (1991-07-01)
A C McWhorter, R L Haddock, F A Nocon, A G Steigerwalt, D J Brenner, S Aleksić, J Bockemühl, J J Farmer
ABSTRAKT
In 1985 the vernacular name Enteric Group 90 was coined for a small group of strains that had been referred to our laboratory as probable strains of Salmonella but did not agglutinate in Salmonella typing antisera. By DNA-DNA hybridization (hydroxyapatite method, 32P), seven strains of Enteric Group 90 were found to be closely related (98 to 100% at 60 degrees C and 94 to 100% at 75 degrees C) to the first strain received (0370-85). The relatedness of Enteric Group 90 to 62 strains of other species of the family Enterobacteriaceae was only 6 to 41%, with the highest values obtained with strains of Salmonella, Kluyvera, Shigella, Klebsiella, Enterobacter, and Citrobacter. We propose a new genus, Trabulsiella, with a single new species, Trabulsiella guamensis, for the highly related group of eight strains formerly known as Enteric Group 90. The type strain is designated ATCC 49490 (CDC 0370-85). T. guamensis strains grew well at 36 degrees C and had positive reactions in the following tests: methyl red, citrate utilization (Simmons) (38% positive at day 1, 88% positive at 2 days), H2S production, lysine decarboxylase, arginine dihydrolase (50% positive at 2 days, 100% positive at 7 days), ornithine decarboxylase, motility, growth in KCN medium, mucate fermentation, acetate utilization, nitrate reduction to nitrite, weak tyrosine hydrolysis (88% positive at 2 days, 100% positive at 7 days), and ONPG (o-nitrophenyl-beta-D-galactopyranoside) test. The strains fermented D-glucose with gas production and fermented L-arabinose, cellobiose, D-galactose, D-galacturonate, maltose, D-mannitol, D-mannose, L-rhamnose, D-sorbitol, trehalose, and D-xylose. T. guamensis strains had negative reactions in the following tests: indole production (13% positive), Voges-Proskauer, urea hydrolysis, phenylalanine deaminase, malonate utilization, lipase (corn oil), DNase, oxidase, pigment production, and acid production from adonitol, D-arabitol, dulcitol, erythritol, myo-inositol, melibiose, alpha-methyl-D-glucoside, raffinose, and sucrose. There were delayed positive reactions for gelatin liquefaction (22 degrees C), which was positive at 12 to 23 days, esculin hydrolysis (13% positive at day 1, 50% positive at 7 days), lactose fermentation (13% positive at 3 to 7 days, 100% positive at 8 to 10 days), glycerol fermentation (88% positive at 7 days), and salicin fermentation (13% positive at day 1, 88% positive at 7 days). All strains were susceptible by the disk diffusion method to colistin, nalidixic acid, gentamicin, streptomycin, kanamycin, chloramphenicol, and trimethoprim-sulfamethoxazole, and most strains were susceptible to sulfadiazine (75% susceptible), tetracycline (88%), and carbenicillin (75%). The strains were resistant to penicillin, cephalothin, and ampicillin. The strains were isolated from vacuum cleaner dust (five strains), soil (one strain), and human feces (two strains). Although T. guamensis can occur in human diarrheal stools, there is no evidence that it actually causes diarrhea. Its main interest to clinical microbiologists may be its possible misidentification as a strain Salmonella.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)