Merck
Antigenic bacterial polysaccharide in rheumatoid synovial effusions.
Arthritis and rheumatism (1979-09-01)
L E Bartholomew, F N Bartholomew
PMID314293
ABSTRAKT
Phenol-water extracted rheumatoid synovial fluids and synovial fluid leukocytes contain an antigen immunologically identical to the Proprionibacterium group bacteria. The antigen was identified by counter-immunoelectrophoresis in 70% of rheumatoid synovial fluid leukocyte pellets and in 60% of rheumatoid synovial fluids. It was also present in 6% of nonrheumatoid fluids and in 22% of nonrheumatoid inflammatory fluid leukocytes. Antigen was not detectable in synovial samples before extraction. Synovial and bacterial antigens were further purified by proteolytic digestion and Sepharose 4B column chromatography. Biochemical and enzymatic studies of bacterial and synovial antigens were similar and consistent with a high molecular weight polysaccharide. Serum antibody to bacterial and synovial antigens was significantly less frequent in rheumatoid sera than in normal controls. The significance of demonstrating a bacterial polysaccharide primarily in rheumatoid synovial effusions is discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)