Merck
Catecholamines, angiotensin II and sodium concentrations in cerebrospinal fluid in young men with borderline hypertension.
Clinical and experimental hypertension. Part A, Theory and practice (1984-01-01)
Y Kawano, K Fukiyama, Y Takeya, I Abe, T Omae
ABSTRAKT
To evaluate the role of central nervous mechanisms and their relationships to the peripheral sympathetic nervous system in borderline hypertension, we measured catecholamines, angiotensin II (AII) and sodium (Na) concentrations in cerebrospinal fluid (CSF) with plasma catecholamines concomitantly in 12 young men with borderline hypertension and 7 age-matched healthy normotensive men on ordinary salt intake. Plasma norepinephrine (NE) and epinephrine (E) were higher in the borderline hypertensives than in the normotensives (NE: 239 +/- 15 vs 190 +/- 11 pg/ml, p less than 0.05, E: 83 +/- 9 vs 43 +/- 6 pg/ml, p less than 0.01). NE levels in CSF were also higher in the borderline hypertensives than in the normotensives (200 +/- 15 vs 150 +/- 18 pg/ml, p less than 0.05). In most of the subjects, CSF E and plasma and CSF dopamine levels were below the sensitivity of the assay. CSF NE correlated positively with both plasma NE (p less than 0.01) and mean blood pressure (p less than 0.05) in all subjects. Immunoreactive AII and Na concentrations in CSF did not differ between the borderline hypertensives and normotensives. These results suggest that peripheral sympathoadrenal overactivity in young subjects with borderline hypertension may be related to an altered function of central noradrenergic neurons. AII and Na in the central nervous system do not appear to have an important role in borderline hypertension.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
(−)-Epinephrine