Merck
[Levels of Krebs cycle metabolites in the blood and urine of patients with diabetes mellitus].
Problemy endokrinologii (1983-03-01)
A S Efimov, M F Gulyĭ, A V Shcherbak, N D Dzvonkevich
ABSTRAKT
The blood oxaloacetate, alpha-ketoglutarate, malate and citrate concentrations and citrate excretion with the urine were studied in 86 patients with diabetes mellitus. An enhanced citrate content and lowered oxaloacetate, malate and alpha-ketoglutarate concentration were detected in the blood of patients with diabetes mellitus. Compensation of the disease and combined pathogenetic therapy are factors, promoting normalization of Krebs cycle metabolite content.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Oxaloacetic acid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
Oxaloacetic acid, powder, suitable for hybridoma
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)