Merck
Identification of CYP4A11 as the major lauric acid omega-hydroxylase in human liver microsomes.
Archives of biochemistry and biophysics (1996-11-01)
P K Powell, I Wolf, J M Lasker
ABSTRAKT
Human liver microsomes are capable of oxidizing lauric acid (laurate), a model medium-chain fatty acid, at both the omega- and omega-1 positions to form 12- and 11-hydroxylaurate, respectively. These laurate hydroxylation reactions are apparently catalyzed by distinct P450 enzymes. While the P450 responsible for microsomal laurate omega-1 hydroxylation in human liver has been identified as CYP2E1, the enzyme catalyzing omega-hydroxylation remains poorly defined. To that end, we employed conventional purification and immunochemical techniques to characterize the major hepatic laurate omega-hydroxylase in humans. Western blotting with rat CYP4A1 antibodies was used to monitor a cross-reactive P450 protein (M(r) = 52 kDa) during its isolation from human liver microsomes. The purified enzyme (7.4 nmol P450/mg protein) had an NH2-terminal amino acid sequence identical to that predicted from the human CYP4A11 cDNA over the first 20 residues found. Upon reconstitution with P450 reductase and cytochrome b5, CYP4A11 proved to be a potent laurate omega-hydroxylase, exhibiting a turnover rate of 45.7 nmol 12-hydroxylaurate formed/min/nmol P450 (12-fold greater than intact microsomes), while catalyzing the omega-1 hydroxylation reaction at much lower rates (5.4 nmol 11-hydroxylaurate formed/min/nmol P450). Analysis of the laurate omega-hydroxylation reaction in human liver microsomes revealed kinetic parameters (a lone Km of 48.9 microM with a VMAX of 3.72 nmol 12-hydroxylaurate formed/min/nmol P450) consistent with catalysis by CYP4A11. In fact, incubation of human liver microsomes with antibodies raised to CYP4A11 resulted in nearly 85% inhibition of laurate omega-hydroxylase activity while omega-1 hydroxylase activity remained unaffected. Furthermore, a strong correlation (r = 0.89; P < 0.001) was found between immunochemically determined CYP4A11 content and laurate omega-hydroxylase activity in liver samples from 11 different subjects. From the foregoing, it appears that CYP4A11 is the principle laurate omega-hydroxylating enzyme expressed in human liver.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecanoic acid, 98%
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)