Merck
Daphnetin prevents chronic unpredictable stress-induced cognitive deficits.
Fundamental & clinical pharmacology (2012-06-22)
Min-Jing Liao, Lian-Feng Lin, Xing Zhou, Xiao-Wen Zhou, Xiao Xu, Xin Cheng, Qin Gao, Huan-Min Luo
ABSTRAKT
Chronic exposure to stress hormones might impair cognitive functions such as learning and memory, which were associated with many mood disorders and neurodegenerative diseases. In this study, we aimed to screen for effective compounds to prevent cognitive deficits induced by chronic stress. Daphnetin was found to protect the cortical neurons against dexamethasone-induced reduction of cell viability in a dose-dependent manner in vitro. We further evaluated its effects on chronic unpredictable stress (CUS) mice in vivo. Two and 8 mg/kg administration of daphnetin could improve the performance of stress mice in Morris water maze tests and forced swimming tests. The results suggested that daphnetin might be a potent compound to treat cognitive deficits induced by CUS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
7,8-Dihydroxycoumarin, ≥97%, powder