Merck
Bonding of simplified adhesive systems to caries-affected dentin of primary teeth.
The journal of adhesive dentistry (2013-03-28)
Fabiana Bucholdz Alves, Tathiane Larissa Lenzi, Alessandra Reis, Alessandro D Loguercio, Thiago Saads Carvalho, Daniela Prócida Raggio
ABSTRAKT
To evaluate the bonding of simplified adhesive systems to sound and caries-affected dentin of primary teeth with microtensile (µTBS) and nanoleakage (NL) tests. Occlusal cavities were prepared in 36 sound second primary molars. Half of the specimens were submitted to pH cycling to simulate caries-affected dentin. Teeth were randomly restored with one of three materials: the etch-and-rinse adhesive system Adper Single Bond 2 (SB), the two-step self-etching adhesive system Adper SE Plus (SE), and the one-step self-etching adhesive system Adper Easy One (EASY). After storage for 24 h, specimens with cross-sectional areas of 0.8 mm2 were prepared for microtensile testing (1 mm/min). One stick from each tooth was immersed in silver nitrate solution (24 h) and allowed to develop for 8 h in order to score the nano leakage with SEM. The fracture pattern was evaluated using a stereomicroscope (400X). The µTBS means were analyzed by two-way ANOVA and Tukey's post-hoc test. For NL, the Kruskal- Wallis and Mann-Whitney tests were used (α < 0.05). SB (35.5 ± 3.5) showed the highest µTBS value to sound dentin, followed by EASY (26.3 ± 1.9) and SE (18.2 ± 6.5) (p < 0.05). No difference among materials was observed for caries-affected dentin (SB: 17.8 ± 4.2; SE: 13.9 ± 3.2; EASY: 14.4 ± 4.2, p > 0.05). For all groups, adhesive/mixed fracture prevailed. Caries affected dentin promoted silver nitrate uptake into the adhesive interface; however, with SE, the nano leakage was more pronounced than in the other adhesive systems, even in sound dentin. Caries-affected dentin negatively influences the bond strength and nano leakage of the two-step etch-and-rinse and one-step self-etching adhesive systems tested in primary teeth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Bisphenol A glycerolate dimethacrylate, glycerol/phenol 1