Merck
Heterologous vaccination against human tuberculosis modulates antigen-specific CD4+ T-cell function.
European journal of immunology (2013-06-06)
One B Dintwe, Cheryl L Day, Erica Smit, Elisa Nemes, Clive Gray, Michele Tameris, Helen McShane, Hassan Mahomed, Willem A Hanekom, Thomas J Scriba
ABSTRAKT
Heterologous prime-boost strategies hold promise for vaccination against tuberculosis. However, the T-cell characteristics required for protection are not known. We proposed that boost vaccines should induce long-lived functional and phenotypic changes to T cells primed by Bacille Calmette Guerin (BCG) and/or natural exposure to mycobacteria. We characterized changes among specific CD4(+) T cells after vaccination with the MVA85A vaccine in adults, adolescents, and children. CD4(+) T cells identified with Ag85A peptide-bearing HLA class II tetramers were characterized by flow cytometry. We also measured proliferative potential and cytokine expression of Ag85A-specific CD4(+) T cells. During the effector phase, MVA85A-induced specific CD4(+) T cells coexpressed IFN-γ and IL-2, skin homing integrins, and the activation marker CD38. This was followed by contraction and a transition to predominantly IL-2-expressing, CD45RA(-) CCR7(+) CD27(+) or CD45RA(+) CCR7(+) CD27(+) specific CD4(+) T cells. These surface phenotypes were similar to Ag85A-specific T cells prior to MVA85A. However, functional differences were observed postvaccination: specific proliferative capacity was markedly higher after 6-12 months than before vaccination. Our data suggest that MVA85A vaccination may modulate Ag85A-specific CD4(+) T-cell function, resulting in greater recall potential. Importantly, surface phenotypes commonly used as proxies for memory T-cell function did not associate with functional effects of vaccination.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
NADase from porcine brain, ≥0.007 unit/mg solid