Merck
Conservative management of postoperative bronchopleural fistulas.
The Journal of thoracic and cardiovascular surgery (2013-07-03)
Mohamed Sadok Boudaya, Hanène Smadhi, Hazem Zribi, Jalel Mohamed, Jamel Ammar, Taher Mestiri, Tarek Kilani
ABSTRAKT
A bronchopleural fistula (BPF) is a serious complication after pulmonary resection and carries a high mortality rate. It remains a therapeutic challenge. The lack of a consensus suggests that no optimal therapy is available; however, endoscopic closure of a fistula may avoid extensive and potentially risky surgery. Seventeen patients (15 men and 2 women) with a BPF after a pneumonectomy (n = 2) or a lobectomy (n = 15), seen between 1995 and 2010, were reviewed. Their median age was 50 years (range, 14-75 years). Underlying diseases were malignant (n = 4) and nonmalignant (n = 13). The mean interval between surgery and fistula development was 20 days (range, 5-270 days). Clinical symptoms leading to a diagnosis of BPF were a persistent air leak (n = 2), a persistent air leak associated with pleural empyema (n = 3), pleural empyema alone (n = 11), and dyspnea (n = 1). Mean fistula size was 3.3 mm (range, 2-9 mm). Treatment consisted of oriented pleural drainage, adequate antibiotic therapy, and endoscopic closure of the fistula with local application of silver nitrate through a flexible bronchoscope (3-15 sessions, 3 times per week). Fistula closure was successful in 16 patients, but failed in 1 patient, who died from acute respiratory distress. BPF is a severe complication in thoracic surgery. The combination of pleural drainage, adequate antibiotic treatment, and mucosal application of silver nitrate, through a flexible bronchoscope, is an efficient alternative and avoids extensive surgical intervention.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh