Merck
Structural and kinetic characterization of two 4-oxalocrotonate tautomerases in Methylibium petroleiphilum strain PM1.
Archives of biochemistry and biophysics (2013-07-09)
Cassidy R Terrell, Elizabeth A Burks, Christian P Whitman, David W Hoffman
ABSTRAKT
Methylibium petroleiphilum strain PM1 uses various petroleum products including the fuel additive methyl tert-butyl ether and straight chain and aromatic hydrocarbons as sole carbon and energy sources. It has two operons, dmpI and dmpII, that code for the enzymes in a pair of parallel meta-fission pathways. In order to understand the roles of the pathways, the 4-oxalocrotonate tautomerase (4-OT) isozyme from each pathway was characterized. Tautomerase I and tautomerase II have the lowest pairwise sequence identity (35%) among the isozyme pairs in the parallel pathways, and could offer insight into substrate preferences and pathway functions. The kinetic parameters of tautomerase I and tautomerase II were determined using 2-hydroxymuconate and 5-(methyl)-2-hydroxymuconate. Both tautomerase I and tautomerase II process the substrates, but with different efficiencies. Crystal structures were determined for both tautomerase I and tautomerase II, at 1.57 and 1.64Å resolution, respectively. The backbones of tautomerase I and tautomerase II are highly similar, but have distinct active site environments. The results, in combination with those for other structurally and kinetically characterized 4-OT isozymes, suggest that tautomerase I catalyzes the tautomerization of both 2-hydroxymuconate and alkyl derivatives, whereas tautomerase II might specialize in other aromatic hydrocarbon metabolites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tetramethylammonium chloride solution, Molecular Biology
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard