Merck
Baseline concentrations of strontium and 90Sr in seawater from the northern Gulf.
Marine pollution bulletin (2013-07-24)
S Uddin, A N Al Ghadban, M Behbahani
ABSTRAKT
Baseline concentration of strontium and Sr-90 in Gulf is presented. The strontium concentration is much higher than reported for other oceanic waters, while the Sr-90 concentration is low at 0.7-1.0 mBq l(-1), that represents the background level following nuclear tests and can be used as an effective tracer in case of any radioactive release in the region. The strontium concentration is primarily related to the increasing salinity off the Gulf coast.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder